Learn More
Mettler Toledo™ Dosing Unit for Excellence and Compact Potentiometric Titrators
Description
- For extended dispensing and titration purposes
- Plug & Play allows easy addition of Dosing Units to the titrator
- Burettes are automatically recognized by the drive
- Connection: CAN
- Resolution (drive): 20'000 steps
Specifications
Specifications
| Connector Types | CAN |
| Type | Dosing Unit |
| For Use With (Application) | Extended dispensing and titration purposes |
| Resolution | 20'000 steps |