missing translation for 'onlineSavingsMsg'
Learn More

Doripenem Monohydrate 98.0+%, TCI America™

Catalog No. D524950MG
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D524950MG 50 mg
D5249250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D524950MG Supplier TCI America Supplier No. D524950MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 364622-82-2
Molecular Formula C15H26N4O7S2
Molecular Weight (g/mol) 438.514
MDL Number MFCD09749887
InChI Key NTUBEBXBDGKBTJ-WGLOMNHJSA-N
PubChem CID 636377
IUPAC Name (4R,5S,6S)-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-3-[(3S,5S)-5-[(sulfamoylamino)methyl]pyrrolidin-3-yl]sulfanyl-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid;hydrate
SMILES CC1C2C(C(=O)N2C(=C1SC3CC(NC3)CNS(=O)(=O)N)C(=O)O)C(C)O.O

Specifications

Melting Point 173°C
Quantity 50 mg
Formula Weight 420.50
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Doripenem Monohydrate
TSCA No
Recommended Storage Refrigerator