Learn More
Thermo Scientific™ Dolly for 300L Plastic Drum for HyPerforma™ Single-Use Mixer DS 300 (stainless steel)
GREENER_CHOICE
Catalog No. SH3095803
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
SH3095803 1 Ea.
1 options
Catalog No. SH3095803 Supplier Thermo Scientific™ Supplier No. SH3095803
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Dolly (single pack) is designed for use with 300L Plastic Drums and the Thermo Scientific™ HyPerforma™ Single-Use Mixer (S.U.M.) DS 300. Dollies for the drums are available in two sizes: one for 50 to 200L drums and another for 300L drums. These drums should be used with the corresponding optional drum positioner that is available with the docking station.
The Dolly (single pack) is designed for use with 300L Plastic Drums and the Thermo Scientific™ HyPerforma™ Single-Use Mixer (S.U.M.) DS 300. Dollies for the drums are available in two sizes: one for 50 to 200L drums and another for 300L drums. These drums should be used with the corresponding optional drum positioner that is available with the docking station.
Required accessories:
  • HyPerforma Single-Use Mixer DS 300 docking station
  • HyPerforma Single-Use Mixer DS 300 bioprocess container bags for closed-top mixing or liners for open-top mixing
Recommended for:
  • Powder-liquid or liquid-liquid mixing applications in single-use
  • High-throughput mixing and storage
Specifications
For Use With (Equipment) DS 300 Single-Use Mixer
Capacity (Metric) 300 L
Product Line HyPerforma
Material Polyethylene
Quantity 1 Ea.