missing translation for 'onlineSavingsMsg'
Learn More

Dodecyltrimethylammonium Chloride 97.0+%, TCI America™

Catalog No. p-7126557
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D101625G 25 g
D1016500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D101625G Supplier TCI America Supplier No. D101625G
Only null left

Chemical Identifiers

CAS 112-00-5
Molecular Formula C15H34ClN
Molecular Weight (g/mol) 263.894
MDL Number MFCD00041974
InChI Key DDXLVDQZPFLQMZ-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium chloride, alicop, cation bb, aliquat 4, n,n,n-trimethyldodecan-1-aminium chloride, laurtrimonium chloride, dehyquart lt, cation fb, nissan cation bb, lauryltrimethylammonium chloride
PubChem CID 8152
IUPAC Name dodecyl(trimethyl)azanium;chloride
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Cl-]

Specifications

Melting Point 246°C
Color White
pH 4.0 to 7.5 (50g/L H2O 25°C)
Quantity 25 g
UN Number 3077
Formula Weight 263.89
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dodecyltrimethylammonium Chloride
EINECSNumber (2)-0184&(9)-1971
RTECSNumber JR2122500
TSCA Yes
Recommended Storage Refrigerator