missing translation for 'onlineSavingsMsg'
Learn More

Dodecyltriethoxysilane 95.0+%, TCI America™

Catalog No. p-7126759
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D151010ML 10 mL
D151025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D151010ML Supplier TCI America Supplier No. D151010ML
Only null left

Chemical Identifiers

CAS 18536-91-9
Molecular Formula C18H40O3Si
Molecular Weight (g/mol) 332.6
MDL Number MFCD00048437
InChI Key YGUFXEJWPRRAEK-UHFFFAOYSA-N
Synonym silane, dodecyltriethoxy, n-dodecyltriethoxysilane, lauryl triethoxy silane, dodecyl triethoxy silane, acmc-209emw, silane,dodecyltriethoxy, dodecyltriethoxysilane, technical
PubChem CID 87696
IUPAC Name dodecyl(triethoxy)silane
SMILES CCCCCCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Color Yellow
Boiling Point 129°C
Quantity 10 mL
Formula Weight 332.60
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecyltriethoxysilane