missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyltriethoxysilane 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 129°C |
| Quantity | 10 mL |
| Formula Weight | 332.60 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dodecyltriethoxysilane |
Safety and Handling
| TSCA | Yes |