missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyltrichlorosilane 97.0+%, TCI America™
Supplier: TCI America D150925ML
Specifications
| Dodecyltrichlorosilane | |
| Yellow | |
| C12H25Cl3Si | |
| 25 mL | |
| dodecyltrichlorosilane, trichloro dodecyl silane, silane, trichlorododecyl, dodecyl trichlorosilane, silane, dodecyltrichloro, n-dodecyltrichlorosilane, dodecyl trichlorosilane dot, hsdb 387, trichlorolaurylsilane, acmc-1ai7g | |
| CCCCCCCCCCCC[Si](Cl)(Cl)Cl | |
| 303.767 | |
| 303.77 | |
| Liquid |
| 4484-72-4 | |
| 294°C | |
| MFCD00000483 | |
| 1771 | |
| BNCXNUWGWUZTCN-UHFFFAOYSA-N | |
| trichloro(dodecyl)silane | |
| 20568 | |
| ≥97.0% (T) |
Chemical Identifiers
| 4484-72-4 | |
| 303.767 | |
| BNCXNUWGWUZTCN-UHFFFAOYSA-N | |
| 20568 | |
| CCCCCCCCCCCC[Si](Cl)(Cl)Cl |
| C12H25Cl3Si | |
| MFCD00000483 | |
| dodecyltrichlorosilane, trichloro dodecyl silane, silane, trichlorododecyl, dodecyl trichlorosilane, silane, dodecyltrichloro, n-dodecyltrichlorosilane, dodecyl trichlorosilane dot, hsdb 387, trichlorolaurylsilane, acmc-1ai7g | |
| trichloro(dodecyl)silane |
Safety and Handling
EINECSNumber : (2)-2041
TSCA : Yes