missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt 98.0+%, TCI America™
Supplier: TCI America D2332250G
Specifications
| Dodecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt | |
| 244°C | |
| C17H37NO3S | |
| 250 g | |
| IZWSFJTYBVKZNK-UHFFFAOYSA-N | |
| 3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate | |
| 84703 | |
| 335.55 | |
| Crystalline Powder |
| 14933-08-5 | |
| White | |
| MFCD00036909 | |
| lauryl sultaine, lauryl sulfobetaine, n-dodecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dodecyldimethylammonio propane-1-sulfonate, zwittergent 3-12, 3-dodecyldimethylammonio propanesulfonate, sulfobetaine 12, unii-r6p3kw3e8u, 3-lauryldimethylammonio propanesulfonate, dodecyldimethyl 3-sulphonatopropyl ammonium | |
| CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] | |
| 335.547 | |
| CHEBI:75303 | |
| ≥98.0% (N) |
Chemical Identifiers
| 14933-08-5 | |
| 335.547 | |
| IZWSFJTYBVKZNK-UHFFFAOYSA-N | |
| 84703 | |
| 3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate |
| C17H37NO3S | |
| MFCD00036909 | |
| lauryl sultaine, lauryl sulfobetaine, n-dodecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dodecyldimethylammonio propane-1-sulfonate, zwittergent 3-12, 3-dodecyldimethylammonio propanesulfonate, sulfobetaine 12, unii-r6p3kw3e8u, 3-lauryldimethylammonio propanesulfonate, dodecyldimethyl 3-sulphonatopropyl ammonium | |
| CHEBI:75303 | |
| CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
Safety and Handling
EINECSNumber : (2)-1664
TSCA : Yes