missing translation for 'onlineSavingsMsg'
Learn More

Dodecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt 98.0+%, TCI America™

Catalog No. D38605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38605G 5 g
D386025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38605G Supplier TCI America Supplier No. D38605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14933-08-5
Molecular Formula C17H37NO3S
Molecular Weight (g/mol) 335.547
MDL Number MFCD00036909
InChI Key IZWSFJTYBVKZNK-UHFFFAOYSA-N
Synonym lauryl sultaine, lauryl sulfobetaine, n-dodecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dodecyldimethylammonio propane-1-sulfonate, zwittergent 3-12, 3-dodecyldimethylammonio propanesulfonate, sulfobetaine 12, unii-r6p3kw3e8u, 3-lauryldimethylammonio propanesulfonate, dodecyldimethyl 3-sulphonatopropyl ammonium
PubChem CID 84703
ChEBI CHEBI:75303
IUPAC Name 3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]

Specifications

Melting Point 244°C
Color White
Quantity 5 g
Formula Weight 335.55
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Dodecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt [for Biochemical Research]