missing translation for 'onlineSavingsMsg'
Learn More

Dodecylbenzene 98.0+%, TCI America™

Catalog No. D107425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D107425ML 25 mL
D1074500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D107425ML Supplier TCI America Supplier No. D107425ML

Chemical Identifiers

CAS 123-01-3
Molecular Formula C18H30
Molecular Weight (g/mol) 246.438
MDL Number MFCD00008974
InChI Key KWKXNDCHNDYVRT-UHFFFAOYSA-N
Synonym 1-phenyldodecane, benzene, dodecyl, n-dodecylbenzene, laurylbenzene, phenyldodecane, marlican, dodecane, 1-phenyl, detergent alkylate, alkylate p 1, phenyldodecan
PubChem CID 31237
IUPAC Name dodecylbenzene
SMILES CCCCCCCCCCCCC1=CC=CC=C1

Specifications

Melting Point 3°C
Color Colorless
Boiling Point 148°C
Quantity 25 mL
Formula Weight 246.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecylbenzene
EINECSNumber (3)-0021
RTECSNumber CZ9540000
TSCA Yes