missing translation for 'onlineSavingsMsg'
Learn More

Dodecylbenzene, mixture of isomers

Catalog No. p-7049928
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3136230 250 g
AA31362A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3136230 Supplier Thermo Scientific Chemicals Supplier No. 03136230
Only null left

CAS: 123-01-3 | C18H30 | 246.438 g/mol

1-Phenyldodecane acts as a pseudo-stationary phase (PSP) marker in the preparation of microemulsion, 1-Phenyldodecane was degraded to phenylacetic acid by an oil-degrading bacterium. it is used to prepare an anionic detergent dodecylbenzene sodium sulfonate. It is a petrochemical-derived substitute for the tallow and coconut oil used in making soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-01-3
Molecular Formula C18H30
Molecular Weight (g/mol) 246.438
MDL Number MFCD00008974
InChI Key KWKXNDCHNDYVRT-UHFFFAOYSA-N
Synonym 1-phenyldodecane, benzene, dodecyl, n-dodecylbenzene, laurylbenzene, phenyldodecane, marlican, dodecane, 1-phenyl, detergent alkylate, alkylate p 1, phenyldodecan
PubChem CID 31237
IUPAC Name dodecylbenzene
SMILES CCCCCCCCCCCCC1=CC=CC=C1

Specifications

Melting Point -3°C
Density 0.87 g/mL
Boiling Point 179°C to 180°C (13 mmHg)
Odor Specific type
Refractive Index 1.49
Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 246.44
Physical Form Liquid
Chemical Name or Material Dodecylbenzene, Mixture of isomers
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only