missing translation for 'onlineSavingsMsg'
Learn More

Dodecylamine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7126737
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D145225G 25 g
D1452100G 100 g
D1452500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D145225G Supplier TCI America Supplier No. D145225G

Chemical Identifiers

CAS 929-73-7
Molecular Formula C12H28ClN
Molecular Weight (g/mol) 221.81
MDL Number MFCD00042017
InChI Key TWFQJFPTTMIETC-UHFFFAOYSA-N
Synonym dodecylamine hydrochloride, laurylamine hydrochloride, n-dodecylamine hydrochloride, dodecan-1-amine hydrochloride, 1-dodecanamine, hydrochloride, dodecylamine, hydrochloride, unii-1v1470j2mf, 1-dodecanamine, hydrochloride 1:1, armeen 12d hydrochloride, acmc-209rjd
PubChem CID 458426
IUPAC Name dodecan-1-aminium chloride
SMILES [Cl-].CCCCCCCCCCCC[NH3+]

Specifications

Color White
Quantity 25 g
Formula Weight 221.81
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dodecylamine Hydrochloride
RTECSNumber JR6750000
TSCA Yes