missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis, Thermo Scientific Chemicals

Catalog No. AC419531000
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC419531000 100 g
AC419530250 25 g
AC419530010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419531000 Supplier Thermo Scientific Chemicals Supplier No. 419531000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry

Specifications

Chemical Name or Material Dodecyl sulfate, sodium salt
Melting Point 206.0°C
Color White
CAS 151-21-3, 7757-82-6, 7647-14-5
Physical Form Fine Crystalline, Flaky or Granular Powder
Quantity 100 g
Flash Point >150°C
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Plastic bottle
Appearance 3% aq. soln. clear, colorless. 1% soln. in EtOH clear, colorless
Assay Percent Range 98%
Molecular Formula C12H25NaO4S
Linear Formula CH3(CH2)11OSO3Na
MDL Number MFCD00036175
Beilstein 01, III, 1786
Fieser 13,281
Merck Index 15, 8768
Synonym sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal
Solubility Information Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C)
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O
IUPAC Name sodium dodecyl sulfate
Molecular Weight (g/mol) 288.38
PubChem CID 3423265
ChEBI CHEBI:8984
Formula Weight 288.38
Percent Purity 98%
Grade Biochemical
Show More Show Less

RUO – Research Use Only