missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyl Isocyanate 97.0+%, TCI America™
Supplier: TCI America D33485G
Specifications
| Dodecyl Isocyanate | |
| Yellow | |
| C13H25NO | |
| 5 g | |
| dodecyl isocyanate, dodecane, 1-isocyanato, dodecylisocyanate, 1-dodecyl isocyanate, n-dodecyl isocyanate, acmc-209jmx, isocyanic acid dodecyl ester | |
| CCCCCCCCCCCCN=C=O | |
| 211.349 | |
| 211.35 | |
| Liquid |
| 4202-38-4 | |
| 161°C | |
| MFCD00041744 | |
| 2206 | |
| YIDSTEJLDQMWBR-UHFFFAOYSA-N | |
| 1-isocyanatododecane | |
| 77873 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 4202-38-4 | |
| 211.349 | |
| YIDSTEJLDQMWBR-UHFFFAOYSA-N | |
| 77873 | |
| CCCCCCCCCCCCN=C=O |
| C13H25NO | |
| MFCD00041744 | |
| dodecyl isocyanate, dodecane, 1-isocyanato, dodecylisocyanate, 1-dodecyl isocyanate, n-dodecyl isocyanate, acmc-209jmx, isocyanic acid dodecyl ester | |
| 1-isocyanatododecane |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator