missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyl Acetate 95.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 180°C |
| Quantity | 500 mL |
| Formula Weight | 228.38 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Dodecyl Acetate |
Safety and Handling
| EINECSNumber | (2)-0735 |
| RTECSNumber | AH3525000 |
| TSCA | Yes |