missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Acetate 95.0+%, TCI America™

Catalog No. A090225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A090225ML 25 mL
A0902500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A090225ML Supplier TCI America Supplier No. A090225ML

Chemical Identifiers

CAS 112-66-3
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.376
MDL Number MFCD00008973
InChI Key VZWGRQBCURJOMT-UHFFFAOYSA-N
Synonym lauryl acetate, acetic acid, dodecyl ester, dodecanol acetate, n-dodecyl acetate, 1-dodecanol acetate, dodecanyl acetate, acetate c-12, n-dodecyl ethanoate, dodecan-1-yl acetate, lauryl ethanoate
PubChem CID 8205
IUPAC Name dodecyl acetate
SMILES CCCCCCCCCCCCOC(=O)C

Specifications

Color Colorless
Boiling Point 180°C
Quantity 25 mL
Formula Weight 228.38
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecyl Acetate
EINECSNumber (2)-0735
RTECSNumber AH3525000
TSCA Yes