missing translation for 'onlineSavingsMsg'
Learn More

Dodecene-1 LAS 98.0+%, TCI America™

Catalog No. D14281G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D14281G Supplier TCI America Supplier No. D14281G

Chemical Identifiers

CAS 25155-30-0
Molecular Formula C18H29NaO3S
Molecular Weight (g/mol) 348.48
MDL Number MFCD00011508
InChI Key HFQQZARZPUDIFP-UHFFFAOYSA-M
Synonym linear-Dodecylbenzenesulfonic Acid Sodium Salt, Sodium linear-Dodecylbenzenesulfonate

Specifications

Color White-Yellow
pH 5.5 - 8.0 (10g/L)
Quantity 1 g
UN Number 3077
Formula Weight 348.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dodecene-1 LAS
EINECSNumber (3)-1884&(3)-1906&(3)-1949
RTECSNumber DB6825000
TSCA Yes