Learn More
Dodecanoyl chloride, 98%
CAS: 112-16-3 | C12H23ClO | 218.77 g/mol
$94.27 - $261.26
Chemical Identifiers
| CAS | 112-16-3 |
|---|---|
| Molecular Formula | C12H23ClO |
| Molecular Weight (g/mol) | 218.77 |
| MDL Number | MFCD00000740 |
| InChI Key | NQGIJDNPUZEBRU-UHFFFAOYSA-N |
| Synonym | lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365 |
| PubChem CID | 8166 |
| IUPAC Name | dodecanoyl chloride |
| SMILES | CCCCCCCCCCCC(Cl)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1329422
|
Thermo Scientific Chemicals
A1329422 |
100 g |
Each for $94.27
|
|
|||||
|
AAA1329436
|
Thermo Scientific Chemicals
A1329436 |
500 g |
Each for $261.26
|
|
|||||
Description
Lauroyl chloride is used as tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals and in the preparation of acylated collagen with water solubility and better surface activity. It is also employed as organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel.It is used in the synthesis of hemicellulose-based hydrophobic biomaterials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLauroyl chloride is used as tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals and in the preparation of acylated collagen with water solubility and better surface activity. It is also employed as organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel.It is used in the synthesis of hemicellulose-based hydrophobic biomaterials.
Solubility
Soluble in ethanol and methanol.
Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.
Chemical Identifiers
| 112-16-3 | |
| 218.77 | |
| NQGIJDNPUZEBRU-UHFFFAOYSA-N | |
| 8166 | |
| CCCCCCCCCCCC(Cl)=O |
| C12H23ClO | |
| MFCD00000740 | |
| lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365 | |
| dodecanoyl chloride |
Specifications
| 112-16-3 | |
| 0.922 | |
| 140°C (284°F) | |
| 1.446 | |
| MFCD00000740 | |
| UN3265 | |
| Moisture sensitive | |
| Soluble in ethanol and methanol. | |
| CCCCCCCCCCCC(Cl)=O | |
| 218.77 | |
| 218.77 | |
| Dodecanoyl chloride |
| -17°C | |
| 130°C to 132°C (7 mmHg) | |
| C12H23ClO | |
| CH3(CH2)10COCl | |
| 100 g | |
| 1281201 | |
| lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365 | |
| NQGIJDNPUZEBRU-UHFFFAOYSA-N | |
| dodecanoyl chloride | |
| 8166 | |
| 98% |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H314-H317-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 203-941-7
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only