missing translation for 'onlineSavingsMsg'
Learn More

Dodecanoyl chloride, 98%

Catalog No. AAA1329422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1329422 100 g
AAA1329436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1329422 Supplier Thermo Scientific Chemicals Supplier No. A1329422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112-16-3 | C12H23ClO | 218.77 g/mol

Lauroyl chloride is used as tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals and in the preparation of acylated collagen with water solubility and better surface activity. It is also employed as organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel.It is used in the synthesis of hemicellulose-based hydrophobic biomaterials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lauroyl chloride is used as tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals and in the preparation of acylated collagen with water solubility and better surface activity. It is also employed as organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel.It is used in the synthesis of hemicellulose-based hydrophobic biomaterials.

Solubility
Soluble in ethanol and methanol.

Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 112-16-3
Molecular Formula C12H23ClO
Molecular Weight (g/mol) 218.77
MDL Number MFCD00000740
InChI Key NQGIJDNPUZEBRU-UHFFFAOYSA-N
Synonym lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365
PubChem CID 8166
IUPAC Name dodecanoyl chloride
SMILES CCCCCCCCCCCC(Cl)=O

Specifications

Melting Point -17°C
Density 0.922
Boiling Point 130°C to 132°C (7 mmHg)
Flash Point 140°C (284°F)
Refractive Index 1.446
Linear Formula CH3(CH2)10COCl
Quantity 100 g
UN Number UN3265
Beilstein 1281201
Sensitivity Moisture sensitive
Solubility Information Soluble in ethanol and methanol.
Formula Weight 218.77
Percent Purity 98%
Chemical Name or Material Dodecanoyl chloride
Show More Show Less
Hazard Category H314-H317-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 203-941-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only