missing translation for 'onlineSavingsMsg'
Learn More

Dodecanophenone 98.0+%, TCI America™

Catalog No. p-7131775
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L001825G 25 g
L0018500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L001825G Supplier TCI America Supplier No. L001825G
Only null left

Chemical Identifiers

CAS 1674-38-0
Molecular Formula C18H28O
Molecular Weight (g/mol) 260.421
MDL Number MFCD00008967
InChI Key DJNJZIFFCJTUDS-UHFFFAOYSA-N
Synonym dodecanophenone, laurophenone, 1-dodecanone, 1-phenyl, phenyl undecyl ketone, 1-phenyl-1-dodecanone, phenyl n-undecyl ketone, n-dodecanophenone, undecyl phenyl ketone, maybridge1_003217
PubChem CID 74292
IUPAC Name 1-phenyldodecan-1-one
SMILES CCCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 205°C
Quantity 25 g
Formula Weight 260.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dodecanophenone