missing translation for 'onlineSavingsMsg'
Learn More

Dodecanophenone, 97%

Catalog No. AAL0201422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0201422 100 g
AAL0201414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0201422 Supplier Thermo Scientific Chemicals Supplier No. L0201422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1674-38-0 | C18H28O | 260.421 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1674-38-0
Molecular Formula C18H28O
Molecular Weight (g/mol) 260.421
MDL Number MFCD00008967
InChI Key DJNJZIFFCJTUDS-UHFFFAOYSA-N
Synonym dodecanophenone, laurophenone, 1-dodecanone, 1-phenyl, phenyl undecyl ketone, 1-phenyl-1-dodecanone, phenyl n-undecyl ketone, n-dodecanophenone, undecyl phenyl ketone, maybridge1_003217
PubChem CID 74292
IUPAC Name 1-phenyldodecan-1-one
SMILES CCCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 44°C to 46°C
Boiling Point 201°C to 202°C (9 mmHg)
Quantity 100 g
Beilstein 2050785
Formula Weight 260.42
Percent Purity 97%
Chemical Name or Material Dodecanophenone
EINECSNumber 216-818-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only