missing translation for 'onlineSavingsMsg'
Learn More

Dodecanoic acid, 98%

CAS: 143-07-7 | C12H24O2 | 200.322 g/mol

Supplier:  Thermo Scientific Chemicals A116720E

 View more versions of this product

Catalog No. AAA116720E


Only null left

Description

Description

Dodecanoic acid is used in the preparation of cosmetics, soaps, alkyd resins and wetting agents. It is also used to measure the molar mass of an unknown substance through freezing point depression. It is also used as a food additive and an active component in a treatment for acne. In addition to this, it is a substrate for acylation of certain proteins based on the murine studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecanoic acid is used in the preparation of cosmetics, soaps, alkyd resins and wetting agents. It is also used to measure the molar mass of an unknown substance through freezing point depression. It is also used as a food additive and an active component in a treatment for acne. In addition to this, it is a substrate for acylation of certain proteins based on the murine studies.

Solubility
Soluble in water, benzene, acetone, alcohol, petroleum ether, dimethyl sulfoxide and dimethyl formamide. Slightly soluble in chloroform.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications

Specifications

Dodecanoic acid
43°C to 45°C
180°C (21 mmHg)
Odorless
MFCD00002736
1099477
lauric acid, n-dodecanoic acid, dodecylic acid, vulvic acid, laurostearic acid, dodecoic acid, duodecylic acid, 1-undecanecarboxylic acid, aliphat no. 4, neo-fat 12
POULHZVOKOAJMA-UHFFFAOYSA-N
dodecanoic acid
3893
200.32
143-07-7
0.883
165°C (329°F)
C12H24O2
2500 g
14,5384
Soluble in water,benzene,acetone,alcohol,petroleum ether,dimethyl sulfoxide and dimethyl formamide. Slightly soluble in chloroform.
CCCCCCCCCCCC(=O)O
200.322
CHEBI:30805
98%

Chemical Identifiers

143-07-7
200.322
POULHZVOKOAJMA-UHFFFAOYSA-N
3893
dodecanoic acid
C12H24O2
MFCD00002736
lauric acid, n-dodecanoic acid, dodecylic acid, vulvic acid, laurostearic acid, dodecoic acid, duodecylic acid, 1-undecanecarboxylic acid, aliphat no. 4, neo-fat 12
CHEBI:30805
CCCCCCCCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P280-P305+P351+P338-P310-P501c

H318

EINECSNumber : 205-582-1

RTECSNumber : OE9800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only