missing translation for 'onlineSavingsMsg'
Learn More

Dodecanedioyl dichloride, 98%, Thermo Scientific™

Chemical Identifiers

CAS 4834-98-4
Molecular Formula C12H20Cl2O2
Molecular Weight (g/mol) 267.2
MDL Number MFCD00012243
InChI Key CNXXEPWXNDFGIG-UHFFFAOYSA-N
Synonym dodecanedioyldichloride, dodecandioyldichloride, dodecanedioyl chloride, acmc-209kct, dodecanedioic acid dichloride, 1,12-dodecanedioyl dichloride, dodecanedioyl dichloride
PubChem CID 2733242
IUPAC Name dodecanedioyl dichloride
SMILES C(CCCCCC(=O)Cl)CCCCC(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC305340050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC305340250
View Documents Promotion Details
N/A 25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4834-98-4
267.2
CNXXEPWXNDFGIG-UHFFFAOYSA-N
2733242
C(CCCCCC(=O)Cl)CCCCC(=O)Cl
C12H20Cl2O2
MFCD00012243
dodecanedioyldichloride, dodecandioyldichloride, dodecanedioyl chloride, acmc-209kct, dodecanedioic acid dichloride, 1,12-dodecanedioyl dichloride, dodecanedioyl dichloride
dodecanedioyl dichloride

Specifications

4834-98-4
140°C (0.5mmHg)
Authentic
C12H20Cl2O2
ClCO(CH2)10COCl
1.06
CNXXEPWXNDFGIG-UHFFFAOYSA-N
dodecanedioyl dichloride
2733242
98%
1.0600g/mL
>110°C
97.5% min. (GC)
1.4670 to 1.469
MFCD00012243
dodecanedioyldichloride, dodecandioyldichloride, dodecanedioyl chloride, acmc-209kct, dodecanedioic acid dichloride, 1,12-dodecanedioyl dichloride, dodecanedioyl dichloride
C(CCCCCC(=O)Cl)CCCCC(=O)Cl
267.2
267.2
Dodecanedioyl dichloride, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

Recommended Storage : Product may darken during storage

SDS
Documents

Documents

RUO – Research Use Only