missing translation for 'onlineSavingsMsg'
Learn More

Dodecanedioyl dichloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 305340050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC305340050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4834-98-4
267.2
CNXXEPWXNDFGIG-UHFFFAOYSA-N
2733242
C(CCCCCC(=O)Cl)CCCCC(=O)Cl
C12H20Cl2O2
MFCD00012243
dodecanedioyldichloride, dodecandioyldichloride, dodecanedioyl chloride, acmc-209kct, dodecanedioic acid dichloride, 1,12-dodecanedioyl dichloride, dodecanedioyl dichloride
dodecanedioyl dichloride

Specifications

Dodecanedioyl dichloride
4834-98-4
140°C (0.5mmHg)
Authentic
Glass bottle
1.4670 to 1.469
5g
1.06
CNXXEPWXNDFGIG-UHFFFAOYSA-N
dodecanedioyl dichloride
2733242
98%
98%
1.0600g/mL
>110°C
97.5% min. (GC)
C12H20Cl2O2
ClCO(CH2)10COCl
MFCD00012243
dodecanedioyldichloride, dodecandioyldichloride, dodecanedioyl chloride, acmc-209kct, dodecanedioic acid dichloride, 1,12-dodecanedioyl dichloride, dodecanedioyl dichloride
C(CCCCCC(=O)Cl)CCCCC(=O)Cl
267.2
267.2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

Recommended Storage : Product may darken during storage

SDS
Documents

Documents

Promotions

Promotions