missing translation for 'onlineSavingsMsg'
Learn More

Dodecanedioic Dihydrazide 98.0+%, TCI America™

Catalog No. p-7127245
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D234225G 25 g
D2342500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D234225G Supplier TCI America Supplier No. D234225G
Only null left

Chemical Identifiers

CAS 4080-98-2
Molecular Formula C12H26N4O2
Molecular Weight (g/mol) 258.366
MDL Number MFCD00043619
InChI Key GRGBENNNGZARRZ-UHFFFAOYSA-N
Synonym Dodecanediohydrazide
PubChem CID 20013
IUPAC Name dodecanedihydrazide
SMILES C(CCCCCC(=O)NN)CCCCC(=O)NN

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 25 g
Formula Weight 258.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dodecanedioic Dihydrazide
EINECSNumber (2)-3246
RTECSNumber JR2400000
TSCA Yes