missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecanedioic Dihydrazide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 190°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 258.37 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dodecanedioic Dihydrazide |
Safety and Handling
| EINECSNumber | (2)-3246 |
| RTECSNumber | JR2400000 |
| TSCA | Yes |