missing translation for 'onlineSavingsMsg'
Learn More

Dodecane 99.0+%, TCI America™

Catalog No. D096825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D096825ML 25 mL
D0968500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D096825ML Supplier TCI America Supplier No. D096825ML

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00008969
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Melting Point -10°C
Color Colorless
Boiling Point 216°C
Quantity 25 mL
Formula Weight 170.34
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecane
EINECSNumber (2)-0010
RTECSNumber JR2125000
TSCA Yes