missing translation for 'onlineSavingsMsg'
Learn More

Dodecane 99.5+%, TCI America™

Catalog No. S02845ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02845ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02845ML Supplier TCI America Supplier No. S02845ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00008969
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Melting Point -12°C
Color Colorless
Boiling Point 216°C
Quantity 5 mL
Formula Weight 170.34
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Dodecane [Standard Material for GC]
EINECSNumber (2)-0010
RTECSNumber JR2125000
TSCA Yes