missing translation for 'onlineSavingsMsg'
Learn More

Dodecane, (n-Dodecane), Reagents

Catalog No. p-200021139
Change view
Click to view available options
Quantity:
100 mL
450 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337274 100 mL
01337273 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337274 Supplier Labchem Inc Supplier No. C2197750100C
Only null left

Dodecane, (n-Dodecane), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-Dodecane, Dihexyl, Bihexyl
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Boiling Range 216°C
Melting Point -9.6°C
Color Colorless
Boiling Point 216°C
Melting Range -9.6°C
Packaging Glass Bottle
Quantity 100 mL
Specific Gravity 0.753
Grade Reagent
Physical Form Liquid
Chemical Name or Material Dodecane, (n-Dodecane)
Show More Show Less