missing translation for 'onlineSavingsMsg'
Learn More

Dodecane, (n-Dodecane), Reagents

Numéro de catalogue. 01337274
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01337274 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01337274 Fournisseur Labchem Inc Code fournisseur C2197750100C
Il en reste null

Dodecane, (n-Dodecane), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Identifiants chimiques

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-Dodecane, Dihexyl, Bihexyl
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Spécifications

Boiling Range 216°C
Melting Point -9.6°C
Color Colorless
Boiling Point 216°C
Melting Range -9.6°C
Packaging Glass Bottle
Quantity 100 mL
Specific Gravity 0.753
Grade Reagent
Physical Form Liquid
Chemical Name or Material Dodecane, (n-Dodecane)
Afficher plus de résultats Afficher moins de résultats