missing translation for 'onlineSavingsMsg'
Learn More

Dodecane, ≥99%, Reagent Grade, Solstice

Catalog No. 60047946
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60047946 100 mL Glass bottle
60047948 500 mL Glass bottle
60047947 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60047946 Supplier Solstice Supplier No. D221104100ML

Reagent Grade,≥99%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00008969
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Melting Point -9.6°C
Color Colorless
Density 0.75g/mL at 25°C
Boiling Point 215°C to 217°C
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3(CH2)10CH3
Quantity 100 mL
Beilstein 1697175
Vapor Density 5.96 (vs air)
Formula Weight 170.33g/mol
Percent Purity ≥99%
Chemical Name or Material Dodecane
Show More Show Less
Hazard Statement P301 + P310-P331
RTECSNumber 203-967-9