missing translation for 'onlineSavingsMsg'
Learn More

Dodecamethylpentasiloxane 98.0+%, TCI America™

Catalog No. D439425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D439425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D439425ML Supplier TCI America Supplier No. D439425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141-63-9
Molecular Formula C12H36O4Si5
Molecular Weight (g/mol) 384.841
MDL Number MFCD00039795
InChI Key FBZANXDWQAVSTQ-UHFFFAOYSA-N
PubChem CID 8853
ChEBI CHEBI:84272
IUPAC Name bis[[dimethyl(trimethylsilyloxy)silyl]oxy]-dimethylsilane
SMILES C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C

Specifications

Melting Point -81°C
Color Colorless
Boiling Point 229°C
Quantity 25 mL
Formula Weight 384.84
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecamethylpentasiloxane
EINECSNumber (7)-0476
RTECSNumber SB0970000
TSCA Yes