missing translation for 'onlineSavingsMsg'
Learn More

Dodecahydrodibenzofuran (mixture of isomers) 98.0+%, TCI America™

Catalog No. p-7128699
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52325G 5 g
D523225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52325G Supplier TCI America Supplier No. D52325G
Only null left

Chemical Identifiers

CAS 13054-98-3
Molecular Formula C12H20O
Molecular Weight (g/mol) 180.29
MDL Number MFCD30749260
InChI Key XTDNMGZRUWMVLT-UHFFFAOYNA-N
PubChem CID 21893810
IUPAC Name 8-oxatricyclo[7.4.0.0²,⁷]tridecane
SMILES C1CCC2C(C1)OC1CCCCC21

Specifications

Color Colorless
Boiling Point 259°C
Quantity 5 g
Formula Weight 180.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecahydrodibenzofuran (mixture of isomers)