missing translation for 'onlineSavingsMsg'
Learn More

Dodecafluorosuberic Acid 97.0+%, TCI America™

$173.37 - $521.94

Chemical Identifiers

CAS 678-45-5
Molecular Formula C8H2F12O4
Molecular Weight (g/mol) 390.08
MDL Number MFCD00153146
InChI Key QYCPVKMFWNBDPV-UHFFFAOYSA-N
Synonym dodecafluorosuberic acid, perfluorosuberic acid, dodecafluorooctanedioic acid, dodecafluoro-1,8-octanedioic acid, perfluorosuberic acid dihydrate, perfluoro-1,8-octanedioic acid, perfluorooctane-1,8-dioic acid, 2,2,3,3,4,4,5,5,6,6,7,7-dodecakis fluoranyl octanedioic acid, dodecafluoro-1,8-octanedioic acid; perfluoro-1,8-octanedioic acid; perfluorosuberic acid
PubChem CID 2733268
IUPAC Name dodecafluorooctanedioic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D24655G
View Documents
TCI America
D24655G
5 g N/A
Only null left
 
D246525G
View Documents
TCI America
D246525G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

678-45-5
390.08
QYCPVKMFWNBDPV-UHFFFAOYSA-N
2733268
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O
C8H2F12O4
MFCD00153146
dodecafluorosuberic acid, perfluorosuberic acid, dodecafluorooctanedioic acid, dodecafluoro-1,8-octanedioic acid, perfluorosuberic acid dihydrate, perfluoro-1,8-octanedioic acid, perfluorooctane-1,8-dioic acid, 2,2,3,3,4,4,5,5,6,6,7,7-dodecakis fluoranyl octanedioic acid, dodecafluoro-1,8-octanedioic acid; perfluoro-1,8-octanedioic acid; perfluorosuberic acid
dodecafluorooctanedioic acid

Specifications

678-45-5
White-Yellow
C8H2F12O4
5 g
QYCPVKMFWNBDPV-UHFFFAOYSA-N
dodecafluorooctanedioic acid
2733268
≥97.0% (T)
Dodecafluorosuberic Acid
143°C
205°C
MFCD00153146
dodecafluorosuberic acid, perfluorosuberic acid, dodecafluorooctanedioic acid, dodecafluoro-1,8-octanedioic acid, perfluorosuberic acid dihydrate, perfluoro-1,8-octanedioic acid, perfluorooctane-1,8-dioic acid, 2,2,3,3,4,4,5,5,6,6,7,7-dodecakis fluoranyl octanedioic acid, dodecafluoro-1,8-octanedioic acid; perfluoro-1,8-octanedioic acid; perfluorosuberic acid
OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O
390.08
390.08
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'