missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecafluorosuberic Acid 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 143°C |
| Color | White-Yellow |
| Boiling Point | 205°C |
| Quantity | 5 g |
| Formula Weight | 390.08 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Dodecafluorosuberic Acid |
Safety and Handling
| TSCA | No |