missing translation for 'onlineSavingsMsg'
Learn More

Dodecacarbonyltriosmium, 99%

Catalog No. p-7049145
Change view
Click to view available options
Quantity:
0.5 g
2 g
Catalog No. Quantity
AA1303802 0.5 g
AA1303804 2 g
2 options

Catalog No. AA1303802

Supplier: Thermo Scientific Chemicals 01303802

Only null left

CAS: 15696-40-9 | C12O12Os3 | 906.81 g/mol

Dodecacarbonyltriosmium is an important precursor to organo-osmium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15696-40-9
Molecular Formula C12O12Os3
Molecular Weight (g/mol) 906.81
MDL Number MFCD00011149
InChI Key VUBLMKVEIPBYME-UHFFFAOYSA-N
Synonym osmium carbonyl, triangulo-dodecacarbonyltriosmium, triosmium dodecacarbonyl, tri-osmium dodecacarbonyl
PubChem CID 6096995
IUPAC Name carbon monoxide;osmium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Os].[Os].[Os]

Specifications

Melting Point 224°C
Quantity 0.5 g
UN Number UN3466
Solubility Information Insoluble in water. Slightly soluble in organic solvents.
Formula Weight 906.73
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Dodecacarbonyltriosmium
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
EINECSNumber 239-789-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only