missing translation for 'onlineSavingsMsg'
Learn More

Dodecacarbonyltetracobalt, 98% min, Thermo Scientific Chemicals

Catalog No. p-7049149
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Catalog No. Quantity
AA1305506 5 g
1 options

Catalog No. AA1305506

Supplier: Thermo Scientific Chemicals 01305506

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17786-31-1
Molecular Formula C12Co4O12
Molecular Weight (g/mol) 571.853
MDL Number MFCD00064698
InChI Key YMFAWOSEDSLYSZ-UHFFFAOYSA-N
Synonym dodecacarbonyltetracobalt, dodecacarbonyltetracobalt min
PubChem CID 6096999
IUPAC Name carbon monoxide;cobalt
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Co].[Co].[Co].[Co]

Specifications

Melting Point 60°C (dec.)
Color Black
Physical Form Crystalline
Quantity 5 g
Linear Formula Co4(CO)12
UN Number UN3179
Formula Weight 571.86
Percent Purity ≤98%
Sensitivity Air Sensitive
Density 2.09g/mL
Chemical Name or Material Dodecacarbonyltetracobalt
Show More Show Less
Hazard Category H228-H301+H331-H317-H350
Hazard Statement GHS H Statement
H228-H301-H331-H351-H317
Flammable solid.
Toxic if swallowed.
Toxic if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P240-P241-P261-P264b-P270-P271-P272-P280g-P281-P301+P310-P302+P352-P304+P340-P308+P313-P311-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.
EINECSNumber 241-763-1
RTECSNumber GG3410000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only