missing translation for 'onlineSavingsMsg'
Learn More

Docosanoic acid, 85%, technical

Catalog No. p-3778489
Change view
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC334812500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC334812500 Supplier Thermo Scientific Chemicals Supplier No. 334812500
Only null left

CAS: 112-85-6 | C22H44O2 | 340.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-85-6
Molecular Formula C22H44O2
Molecular Weight (g/mol) 340.58
MDL Number MFCD00002807
InChI Key UKMSUNONTOPOIO-UHFFFAOYSA-N
Synonym behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure
PubChem CID 8215
ChEBI CHEBI:28941
IUPAC Name docosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 80.0°C to 82.0°C
Color White to Yellow
Boiling Point 306.0°C (60.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Packaging Glass bottle
Quantity 250 g
Beilstein 02, 391
Merck Index 15, 1022
Formula Weight 340.58
Percent Purity 85%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material Docosanoic acid
Show More Show Less
EINECSNumber 204-010-8

RUO – Research Use Only