missing translation for 'onlineSavingsMsg'
Learn More

Docetaxel 98.0+%, TCI America™

Catalog No. p-7128098
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4102100MG 100 mg
D41021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4102100MG Supplier TCI America Supplier No. D4102100MG

Chemical Identifiers

CAS 114977-28-5
Molecular Formula C43H53NO14
Molecular Weight (g/mol) 807.89
MDL Number MFCD00800737
InChI Key ZDZOTLJHXYCWBA-VCVYQWHSSA-N
Synonym docetaxel, taxotere, docetaxel anhydrous, docetaxol, emdoc, docetaxel, trihydrate, docetaxel inn, taxotere tn, unii-699121phca, txl
PubChem CID 148124
ChEBI CHEBI:4672
SMILES CC1=C2C(C(=O)C3(C(CC4C(C3C(C(C2(C)C)(CC1OC(=O)C(C(C5=CC=CC=C5)NC(=O)OC(C)(C)C)O)O)OC(=O)C6=CC=CC=C6)(CO4)OC(=O)C)O)C)O

Specifications

Melting Point 188°C
Color White
Quantity 100 mg
Formula Weight 807.89
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Docetaxel
RTECSNumber DA4172750
TSCA No