Learn More
MP Biomedicals™ dNTPs Mix (10mm each)
Improves PCR efficiency, lowers denaturation temperatures and increases template lengths. MP Biomedicals dNTPs Mix (10mm each) is a ready to use solution with guaranteed purification of dNTPs (>99% pure) in order to achieve sensitive and consistent res
Supplier: MP Biomedicals™ 11NTPMX100
Description
Manufacturing process guarantees that the dNTPs are highly purified, as contaminants can hinder PCR and result in a disappointing product yield. This product achieves sensitive and consistent results with specially prepared deoxynucleotides at a concentration of 10mm.
It is suitable for use in a range of molecular biology applications, including: PCR, real-time PCR, high fidelity and long PCR, RT-PCR, cDNA synthesis, and DNA sequencing and labeling procedures.
Specifications
| 10 mM | |
| 7 | |
| Standard PCR, Real Time PCR (qPCR), RT-PCR |
| Store at -20°C. | |
| 5 μmol |