missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNase/RNase-Free Deionized Water, 1 L

Catalog No. A57775
Change view
Click to view available options
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A57775 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A57775 Supplier Thermo Scientific™ Supplier No. A57775

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

DNase/RNase-Free Deionized Water for use in all molecular biology applications.

DNase/RNase-Free Deionized Water is designed for use in all molecular biology applications and provided in a convenient 1-liter and 2-liter bottles.

  • There is no detectable DNase, RNase, or protease activity.
  • It is 0.1-μm membrane-filtered and tested for DNase and RNase activity.

Specifications

Synonym Nuclease-free deionized water
Grade Molecular Biology
Treatment(s) Not DEPC-Treated
Quantity 1 L

For Research Use Only.