Learn More
Stemcell Technologies DNase I Solution (1 mg/mL), Size: 1 mL DFS Item
Supplier:  Stemcell Technologies Inc 07900
Deoxyribonuclease (DNase) I Solution (1 mg/mL) is useful to reduce or prevent the clumping of concentrated and/or cryopreserved cell suspensions following thawing. The activity range is 2000 units per mg of protein.
Catalog No. NC9007308
Only null left
Explore available promotions