Learn More
RayBiotech Inc DNA Purification Magnetic Beads
Catalog No. NC1838875
Encompass
Paramagnetic beads with uniform particle size efficiently bind DNA, resulting in high yields of DNA with minimal RNA, proteins, nucleases, and other cellular contaminants.
Catalog No. NC1838875 Supplier RayBiotech Inc Supplier No. 801109
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Paramagnetic beads with uniform particle size efficiently bind DNA, resulting in high yields of DNA with minimal RNA, proteins, nucleases, and other cellular contaminants.