Learn More
DNA Molecular Model Kit
Catalog No. S65372
Encompass
Change view
Click to view available options
Product Type:
DNA Molecular Model Kit
Catalog No. Product Type
S65372 DNA Molecular Model Kit
1 options
Catalog No. S65372 Supplier Ryler Enterprises Inc Supplier No. DNA1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
a lab activity, a student worksheet, and the pieces listed below.
Construct your DNA!
Assemble a 12 base pair double helix of DNA with this kit. Form hydrogen bonds, twist the model into a helix, and see the complementary DNA bases.
Specifications
For Use With (Application) Used to show a variety of base sequences and the point mutation that causes sickle cell anemia
Includes Lab activity, a student worksheet, 12 base pair double helix of DNA
Product Type DNA Molecular Model Kit