Learn More
Diagenode Inc DNA Methylation control package
Catalog No. NC1341363
Encompass
The DNA Methylation control package includes unmethylated and in vitro methylated DNA together with specific primer sets for assessing the efficiency of your non-plant MeDIP experiments (Methylated DNA Immunoprecipitation) carried out with Diagenodes MagMeDIP qPCR and Auto MagMeDIP qPCR Kit. These controls are made from A. thaliana BAC.
Catalog No. NC1341363 Supplier Diagenode Inc Supplier No. C02040012
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The DNA Methylation control package includes unmethylated and in vitro methylated DNA together with specific primer sets for assessing the efficiency of your non-plant MeDIP experiments (Methylated DNA Immunoprecipitation) carried out with Diagenodes MagMeDIP qPCR and Auto MagMeDIP qPCR Kit. These controls are made from A. thaliana BAC.