missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ DNA LoBind™ Tubes
GREENER_CHOICE

Catalog No. 05414202
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2.0 mL
5 mL
25 mL
50 mL
Packaging:
1 Bag x 500 Tubes
200 Tubes (2 Bags x 100 Tubes)
200 Tubes (4 Bags x 50 Tubes)
200 Tubes (5 Bags x 40 Tubes)
200 Tubes (8 Bags x 25 Tubes)
4 Bags of 50
Closure Type:
Screw Cap
Snap Cap
SnapTec Cap
Max. RCF:
17,000 x g
19,500 x g
25,000 x g
30,000 x g
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Packaging Closure Type Max. RCF
05414202 0.5 mL 1 Bag x 500 Tubes Snap Cap 30,000 x g
05414203 1.5 mL 1 Bag x 500 Tubes Snap Cap 30,000 x g
05414204 2.0 mL 1 Bag x 500 Tubes Snap Cap 25,000 x g
05413916 5 mL 200 Tubes (2 Bags x 100 Tubes) Screw Cap 25,000 x g
01670416 25 mL 200 Tubes (4 Bags x 50 Tubes) Screw Cap 17,000 x g
01670414 25 mL 200 Tubes (5 Bags x 40 Tubes) SnapTec Cap 17,000 x g
361042553 50 mL 200 Tubes (8 Bags x 25 Tubes) Screw Cap 19,500 x g
14282305 5 mL 4 Bags of 50 Snap Cap 25,000 x g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 05414202 Supplier Eppendorf™ Supplier No. 0030108400
Only null left

Unleash the power of precision in your DNA research with Eppendorf DNA LoBind Tubes. Engineered with proprietary LoBind technology, these microcentrifuge tubes redefine sample handling, minimizing the risk of loss and maximizing DNA recovery. Ideal for low-concentration applications, including PCR, qPCR, and DNA sequencing, these tubes ensure accuracy, purity, and optimal yields.

Maximize sample recovery of nucleic acids by significantly reducing sample-to-surface binding with these tubes. Eppendorf™ Polypropylene DNA LoBind Tubes are ideal for sample preparation and long-term storage.

  • Surface-coating-free, including substances like silicone, to reduce the risk of sample interference.
  • Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors.
  • Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments.
  • Precision lid sealing minimizes evaporation rates in the tube format.
  • Suitable for molecular biology applications with a centrifugation speed rating of up to 30,000 x g (25,000 x g for 2.0 mL tube).
  • Snap, screw, and SnapTec™ cap options available.

Specifications

Closure Material HDPE
Color Clear
Capacity (Metric) 0.5 mL
Material Polypropylene
Packaging 1 Bag x 500 Tubes
Quantity 1 Bag of 500 Tubes
Sterility Non-sterile
Autoclavable Autoclavable
Shape Cylindrical
Clean Claims PCR Clean
Bottom Shape Conical
Closure Type Snap Cap
Product Type Tube
Max. RCF 30,000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Show More Show Less