Learn More
Biotium DMSO, anhydrous
Supplier: Biotium 90082/DUP
Anhydrous DMSO (dimethyl sulfoxide; methyl sulfoxide) is recommended for preparing stock solutions of AM ester dyes, reactive dyes and other related moisture-sensitive products. Properties: Colorless liquid at room temperature; Store desiccated at room temperature; C2H6OS; MW: 78.13; [67-68-5].