Learn More
Leica Microsystems DMi1 Manual Inverted Microscope
The Leica DMi1 is a high-quality manual inverted microscope, perfect for cell culture labs. It offers exceptional clarity, precision optics, and user-friendly features, making it ideal for observing cell cultures and individual cells. Elevate your research with its reliable imaging capabilities.
Supplier:  Leica Microsystems 8122660
Catalog No. 01672695
Only null left
Description
Description
The Leica DMi1 for Cell Culture is a robust manual inverted microscope designed for routine tasks in cell culture labs. With precision optics and user-friendly features, this microscope delivers exceptional clarity and detail, making it an ideal tool for observing cell cultures and individual cells. Elevate your cell culture research with the Leica DMI1's outstanding performance and reliable imaging capabilities.
  • Leica DMi1 FOV20 Basic stand FOV20, LED illumination, exchangeable eyepieces, Fix stage, power cable
  • Two eyepieces(10X/20 eyepiece w/eyeguard and 10X/20 focusing eyepiece w/eyeguard)
  • S80 condenser
  • 4 position slider for S80 condenser(BF,Ph0,Ph1,Ph2)
  • 3 Objectives(HI PLAN 4x/0.10 PH0, HI PLAN I 10x/0.22 PH1, HI PLAN I 20x/0.30 PH1)
  • Dust cover
Includes:
  • 11523231 DMi1 FOV20, S80, 4P Slider, 3Obj. IVD
Specifications
Specifications
100/240 V
Plan Achromatic
LED
None
DMi1
40X, 100X, 200X
10X/20 mm
Binocular
None
Educational Microscope
Product Suggestions
Product Suggestions