missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ DMEM/F-12 PLUS Basal Medium

Catalog No. SCM162MI
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM162MI 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM162MI Supplier MilliporeSigma™ Supplier No. SCM162

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Advanced DMEMF12 basal media used in low-serum or serum-free media formulations

  • Proprietary formulation that contains human serum albumin (HSA), defined lipids, trace elements, chemicals and proteins essential for optimal cell growth in reduced serum conditions
  • Media contains high-glucose and does not contain L-glutamine or penicillin-streptomycin
  • No growth/Pass
  • Uses:
    • Low-serum or serum-free media formulations including stem cell and 3D organoid cell cultures
    • Culture of primary, stem cells and cell lines in applications that require reduced or serum-free medium
    • Optimization of transfections that require reduced or serum-free conditions
    • Production of conditioned medium in reduced serum or serum-free conditions
    • Optimized media for the culture and expansion of multiple 3D organoids and tissues systems
  • Specifications

    Form Liquid
    For Use With (Application) Cell Culture, Stem Cell Culture
    Quantity 500 mL
    Endotoxin Level <2 EU/mL
    Osmolality 290 to 330 mOsm
    pH 7.2 to 7.4
    Color Red
    Content And Storage Store 2°C to 8°C, Aliquot into smaller volumes, Protect from light, Acclimate medium room temp 15 to 20 min before use, Do not warm media (at 37 °C before use)

    For research purposes only