missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Valinol 98.0+%, TCI America™
$70.84 - $281.48
Chemical Identifiers
| CAS | 16369-05-4 |
|---|---|
| Molecular Formula | C5H13NO |
| Molecular Weight (g/mol) | 103.165 |
| MDL Number | MFCD00004730 |
| InChI Key | NWYYWIJOWOLJNR-UHFFFAOYSA-N |
| Synonym | 2-amino-3-methyl-1-butanol, dl-valinol, valinol, l-+-valinol, 2-amino-3-methyl-butan-1-ol, 1-butanol, 2-amino-3-methyl, 1-butanol, 2-amino-3-methyl-, .+/-., s-+-2-amino-3-methyl-1-butanol, 1-2-amino-3-methylbutan-1-ol |
| PubChem CID | 79019 |
| IUPAC Name | 2-amino-3-methylbutan-1-ol |
| SMILES | CC(C)C(CO)N |
Chemical Identifiers
| 16369-05-4 | |
| 103.165 | |
| NWYYWIJOWOLJNR-UHFFFAOYSA-N | |
| 79019 | |
| CC(C)C(CO)N |
| C5H13NO | |
| MFCD00004730 | |
| 2-amino-3-methyl-1-butanol, dl-valinol, valinol, l-+-valinol, 2-amino-3-methyl-butan-1-ol, 1-butanol, 2-amino-3-methyl, 1-butanol, 2-amino-3-methyl-, .+/-., s-+-2-amino-3-methyl-1-butanol, 1-2-amino-3-methylbutan-1-ol | |
| 2-amino-3-methylbutan-1-ol |
Specifications
| 16369-05-4 | |
| 87°C | |
| MFCD00004730 | |
| 2-amino-3-methyl-1-butanol, dl-valinol, valinol, l-+-valinol, 2-amino-3-methyl-butan-1-ol, 1-butanol, 2-amino-3-methyl, 1-butanol, 2-amino-3-methyl-, .+/-., s-+-2-amino-3-methyl-1-butanol, 1-2-amino-3-methylbutan-1-ol | |
| CC(C)C(CO)N | |
| 103.165 | |
| 103.17 | |
| Liquid |
| Yellow | |
| C5H13NO | |
| 1 g | |
| NWYYWIJOWOLJNR-UHFFFAOYSA-N | |
| 2-amino-3-methylbutan-1-ol | |
| 79019 | |
| ≥98.0% (GC,T) | |
| DL-Valinol |
Safety and Handling
TSCA : No